Tisk…
Human recombinant ST8SIA1 (from cells)
Dodavatel: ProSci Inc.
91-569
PRSI91-569EA 24260 CZK
PRSI91-569
Human recombinant ST8SIA1 (from cells)
Proteiny
alpha-N-Acetylneuraminide alpha-2,8-Sialyltransferase (ST8SIA1) belongs to the glycosyltransferase 29 family. ST8SIA1 is a sialytransferase that catalyzes the transfer of sialic acid from CMP-sialic acid to GM3 to produce GD3 and GT3. ST8SIA1 is highly expressed in melanoma cell lines, adult and fetal brain, low expressed in adult and fetal lung. ST8SIA1 may act as a type II transmembrane protein with a short N-termianl cytoplasmic domain and a single-pass transmembrane domain folowed by an enzymatic domain in the lument of the Golgi apparatus.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag