Tisk…
Human recombinant Triosephosphate isomerase (from E. coli)
Dodavatel: ProSci Inc.
91-240
PRSI91-240EA 25170 CZK
PRSI91-240
Human recombinant Triosephosphate isomerase (from E. coli)
Enzymy
Triose-phosphate isomerase, also named Triose-phosphate isomerase, TPI and TIM, is an enzyme that catalyses the reversible interconversion of the triose phosphate isomers dihydroxyacetone phosphate and D-glyceraldehyde 3-phosphate. TPI has been found in nearly every organism searched for the enzyme, including animals such as mammals and insects as well as in fungi, plants, and bacteria. However, some bacteria that do not perform glycolysis, like ureaplasmas, lack TPI. TPI plays an important role in glycolysis and is essential for efficient energy production. TPI deficiency is an autosomal recessive disorder and the most severe clinical disorder of glycolysis. Triose phosphate isomerase deficiency is associated with neonatal jaundice, chronic hemolytic anemia, progressive neuromuscular dysfunction, cardiomyopathy and increased susceptibility to infection and characterised by chronic hemolytic anemia.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Objednat
Produktová specifikace
Název enzymu Triosephosphate Isomerase
Environmentally Preferable
Typ enzymu Recombinant
Zdroj E. coli
Druhy Human
Sekvence Met1-Gln249
Molekulární hmotnost 28.8 kD
Čistota >95% as determined by reducing SDS-PAGE
Podmínky uskladnění Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
Formulace Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 1 mM DTT, 10% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Sekvence značek N-6 His tag