Tisk…
Mouse recombinant TIM4 (from CHO cells)
Dodavatel: ProSci Inc.
90-552
PRSI90-552EA 22010 CZK
PRSI90-552
Mouse recombinant TIM4 (from CHO cells)
Proteiny
Mouse T cell immunoglobulin and mucin domain-containing protein 4, also known as T cell membrane protein 4, TIMD4 and TIM4 is a single-pass type I membrane protein which belongs to the immunoglobulin superfamily and TIM family. TIM4 contains one Ig-like V-type (immunoglobulin-like) domain. It is expressed on dendritic cells and macrophages. TIM4 plays an important role in the proliferation of T helper type 2 (Th2) cells. TIM4 is involved in regulating T cell proliferation and lymphotoxin signalling. It is a ligand for HAVCR1 / TIMD1. The expression of TIM4 in fibroblasts enhanced their ability to engulf apoptotic cells. TIM4 and TIM1 are phosphatidylserine receptors for the engulfment of apoptotic cells and may also be involved in intercellular signalling in which exosomes are involved. Modulation of TIM4 production in dendritic cells (DCs) represents a novel therapeutic approach for the treatment of peanut allergy.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.