Tisk…
Anti-Tcrb Hamster Monoclonal Antibody [clone: H57-597] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Dodavatel: ProSci Inc.
77-213
PRSI77-213EA 13900 CZK
PRSI77-213
Anti-Tcrb Hamster Monoclonal Antibody [clone: H57-597] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Protilátky
The H57-597 monoclonal antibody is specific for the beta chain of the mouse T cell Receptor (TCR). The crosslinking induces activation and proliferation of T cells, as a plate-bound or a soluble H57-597, or the plate-bound antibody can induce apoptosis, based on assay conditions. The beta chain of the TCR can combine with the alpha chain of the receptor to produce the alpha-beta TCR, which is expressed by the NKT cells, by the NK1.1+ thymocytes and most of the T cells. The beta chain does not react with the gamma-delta TCR-bearing cells, expressed by a small number of T cells. This antibodyMore Product Information