Tisk…
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (FITC (Fluorescein Isothiocyanate))
Dodavatel: ProSci Inc.
76-414
PRSI76-414EA 9000 CZK
PRSI76-414
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (FITC (Fluorescein Isothiocyanate))
Protilátky
The IM7 monoclonal antibody specifically reacts with all the isoforms and both alloantigens of the CD44 glycoprotein (Pgp-1, Ly-24). CD44 is expressed on hematopoietic and non-hematopoietic cells, bone marrow myeloid cells, memory T lymphocytes, periphery activated B cells, CD4+ T lymphocytes, and CD8+ T lymphocytes. The periphery B and T lymphocytes upregulate the expression of CD44. CD44 binds to hyaluronan molecules, acting as an adhesion molecule.The IM7 antibody inhibits collagen-induced arthritis in DBA/1 mice, prevents central nervous system inflammation associated with experimentalMore Product Information