Tisk…
Anti-GLS2 Rabbit Polyclonal Antibody
Dodavatel: ProSci Inc.
6217
PRSI6217EA 12390 CZK
PRSI6217
Anti-GLS2 Rabbit Polyclonal Antibody
Protilátky
GLS2 Antibody: Phosphate-activated glutaminase, also known as Glutaminase 2 (GLS2), was initially isolated from rat liver, although it has been shown to be expressed in other tissues. Like the functionally similar, larger kidney glutaminase, GLS2 catalyzes the hydrolysis of glutamine to stoichiometric amounts of glutamate and ammonia. Expression of GLS2 is increased by p53 under both stressed and nonstressed conditions, resulting in increased levels of glutamate and alpha-ketoglutarate, which in turn results in enhanced mitochondrial respiration and ATP generation. GLS2 also regulatesMore Product Information