Tisk…

Anti-ERVW-1 Rabbit Polyclonal Antibody

Dodavatel: ProSci Inc.
58-894
PRSI58-894EA 17660 CZK
PRSI58-894
Anti-ERVW-1 Rabbit Polyclonal Antibody
Protilátky
For WB starting dilution is: 1:1000.

Type: Primary
Antigen: ERVW-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 493-521
Host: Rabbit
Isotype: Ig
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR