Tisk…

Anti-RPL12 Rabbit Polyclonal Antibody

Dodavatel: ProSci Inc.
58-375
PRSI58-375EA 17660 CZK
PRSI58-375
Anti-RPL12 Rabbit Polyclonal Antibody
Protilátky
For WB starting dilution is: 1:1000.

Type: Primary
Antigen: RPL12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 71-100
Host: Rabbit
Isotype: Ig
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR