Tisk…
Anti-CDH19 Rabbit Polyclonal Antibody
Dodavatel: ProSci Inc.
56-755
PRSI56-755EA 17660 CZK
PRSI56-755
Anti-CDH19 Rabbit Polyclonal Antibody
Protilátky
This gene is a type II classical cadherin from the cadherin superfamily and one of three cadherin 7-like genes located in a cluster on chromosome 18. The encoded membrane protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. Since disturbance of intracellular adhesion is a prerequisite for invasion and metastasis of tumor cells, cadherinsMore Product Information