Tisk…
Anti-PPAPDC1A Rabbit Polyclonal Antibody
Dodavatel: ProSci Inc.
5513
PRSI5513EA 12390 CZK
PRSI5513
Anti-PPAPDC1A Rabbit Polyclonal Antibody
Protilátky
PPAPDC1A Antibody: Phosphatidate phosphatase (PAP) plays important role in lipid-signaling metabolism in eukaryotic cells. Two distinct types of PAP (PAP1 and PAP2) activity have been distinguished by their subcellular localization and differential sensitivity to N-ethylmaleimide(NEM) and Mg2+. A yeast diacylglycerol pyrophosphate (DGPP) phosphatase (DPP1) and mammalian DGPP phosphatase (PAP2) have been identified as Mg2+-independent and NEM-insensitive membrane-associated. PPAPDC1A (also known as DPPL2) and PPAPDC1B (DPPL1) form a novel type of Mg2+-independent and NEM-sensitive mammalianMore Product Information
Objednat
Produktová specifikace
Symbol antigenu PPAPDC1A
Název antigenu Phosphatidic Acid Phosphatase Type 2 Domain-Containing Protein 1A
Environmentally Preferable
Synonyma antigenu Phosphatidate phosphatase PPAPDC1A|Phosphatidic acid phosphatase type 2 domain-containing protein 1A|PPAPDC1|PPAPDC1A|DPPL2
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Izotyp IgG
Western blot Yes
ELISA Yes
ImunoFluorescence Yes
Imunogen PPAPDC1A antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human PPAPDC1A.
Čištění Affinity chromatography purified via peptide column
Skladovací pufr PBS containing 0.02% sodium azide
Testované aplikace E, WB, IF