Tisk…
Anti-GNAT3 Rabbit Polyclonal Antibody
Dodavatel: ProSci Inc.
55-210
PRSI55-210EA 17660 CZK
PRSI55-210
Anti-GNAT3 Rabbit Polyclonal Antibody
Protilátky
Guanine nucleotide-binding protein (G protein) alpha subunit playing a prominent role in bitter and sweet taste transduction as well as in umami (monosodium glutamate, monopotassium glutamate, and inosine monophosphate) taste transduction. Transduction by this alpha subunit involves coupling of specific cell-surface receptors with a cGMP-phosphodiesterase; Activation of phosphodiesterase lowers intracellular levels of cAMP and cGMP which may open a cyclic nucleotide-suppressible cation channel leading to influx of calcium, ultimately leading to release of neurotransmitter. Indeed, denatoniumMore Product Information
New Product
Objednat
Produktová specifikace
Symbol antigenu GNAT3
Název antigenu Guanine nucleotide-binding protein G(t) subunit alpha-3
Environmentally Preferable
Synonyma antigenu GNAT3
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human
Hostitel Rabbit
Identifikační číslo genu A8MTJ3
Izotyp
Western blot Yes
Imunochemie Yes
Průtoková cytometrie Yes
Modifikace None
Epitop Amino Acid Range = 78-105
Imunogen This GNAT3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 78-105 amino acids from the Central region of human GNAT3.
Formulář Liquid
Molekulární hmotnost 40 kDa
Čištění This antibody is purified through a protein A column, followed by peptide affinity purification.
Koncentrace 0,5 mg/ml
Skladovací pufr Supplied in PBS with 0,09% (W/V) sodium azide
Skladovací teplota Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Transportní teplota 4 °C