Tisk…
Anti-CRTC1 Rabbit Polyclonal Antibody
Dodavatel: ProSci Inc.
49-411
PRSI49-411EA 20900 CZK
PRSI49-411
Anti-CRTC1 Rabbit Polyclonal Antibody
Protilátky
MECT1 (also known as MucoEpidermoid Carcinoma Translocated 1, Transducer of regulated cAMP response element-binding protein 1, TORC1, and Transducer of CREB protein 1) is a nuclear protein that functions as a transcriptional coactivator for CREB1, which activates transcription through both consensus and variant cAMP response element (CRE) sites. MECT1does not appear to modulate CREB1 DNA-binding activity but enhances the interaction of CREB1 with TAF4/TAFII-130. MECT1 translocates with MAML2 (MasterMind-Like Protein 2) to yield a fusion oncogene: t(11;19) (q21;p13). This translocation occursMore Product Information