Tisk…

Anti-STS Goat Polyclonal Antibody

Dodavatel: ProSci Inc.
45-297
PRSI45-297EA 17660 CZK
PRSI45-297
Anti-STS Goat Polyclonal Antibody
Protilátky
Arylsulfatase C antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Arylsulfatase C antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: STS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR