Tisk…

Anti-Aspergillus antigen Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: ProSci Inc.
35-107
PRSI35-107EA 16750 CZK
PRSI35-107
Anti-Aspergillus antigen Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Purified IgG fraction of antiserum covalently coupled with highly purified preparation of horseradish peroxidase (RZ>3). Care is taken to insure adequate conjugation while preserving the maximum enzyme activity. Free enzyme is absent.

  • Aspergillus sp. antibody is in a phosphate saline buffer with BSA (10mg/ml) and preserved with 0.002% thimerosal.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: Aspergillus antigen
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Fungi
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR