Tisk…
Anti-EPCAM Mouse Monoclonal Antibody [clone: EPM17-2]
Dodavatel: ProSci Inc.
34-061
PRSI34-061EA 22010 CZK
PRSI34-061
Anti-EPCAM Mouse Monoclonal Antibody [clone: EPM17-2]
Protilátky
EpCAM may act as a physical homophilic interaction molecule between intestinal epithelial cells (IECs) and intraepithelial lymphocytes (IELs) at the mucosal epithelium for providing immunological barrier as a first line of defense against mucosal infection. [UniProt].
Western blot: 1 - 2 µg/ml Flow Cytometry: 0,5 - 1 µg/10^6 cells IF: 1 - 2 µg/ml IHC (FFPE): 0,5 - 1 µg/ml for 30 min at RT (1) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the anti-EpCAM antibody to be titered up or down for optimalMore Product Information