Tisk…
Anti-SOX10 Mouse Monoclonal Antibody [clone: SPM607]
Dodavatel: ProSci Inc.
33-464
PRSI33-464EA 22010 CZK
PRSI33-464
Anti-SOX10 Mouse Monoclonal Antibody [clone: SPM607]
Protilátky
Transcription factor that seems to function synergistically with the POU domain protein TST-1/OCT6/SCIP. Could confer cell specificity to the function of other transcription factors in developing and mature glia (By similarity). [UniProt].
Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 1 - 2 µg/ml Western blot: 0,5 - 1 µg/ml Immunohistochemistry (FFPE): 0,5 - 1 µg/ml for 30 min at RT (1) Optimal dilution of the SOX10 antibody should be determined by the researcher.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM Citrate buffer, pH 6,0,
More Product Information