Tisk…

Anti-PTN Rabbit Polyclonal Antibody

Dodavatel: ProSci Inc.
30-059
PRSI30-059EA 18800 CZK
PRSI30-059
Anti-PTN Rabbit Polyclonal Antibody
Protilátky
PTN is a heparin binding mitogenic protein. It has neurite extension activity.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PTN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Objednat

Produktová specifikace

Symbol antigenu PTN
Název antigenu Pleiotrophin
Environmentally Preferable
Synonyma antigenu NEGF1|HBNF|HARP|HBGF8
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Izotyp
Western blot Yes
ELISA Yes
Imunogen A synthetic peptide corresponding a region of human PTN.
Formulář Lyophilized

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR