Tisk…
Anti-SF3A3 Rabbit Polyclonal Antibody
Dodavatel: ProSci Inc.
29-393
PRSI29-393EA 18800 CZK
PRSI29-393
Anti-SF3A3 Rabbit Polyclonal Antibody
Protilátky
SF3A3 is the subunit 3 of the splicing factor 3a protein complex. The splicing factor 3a heterotrimer includes subunits 1, 2 and 3 and is necessary for the in vitro conversion of 15S U2 snRNP into an active 17S particle that performs pre-mRNA splicing. Subunit 3 interacts with subunit 1 through its amino-terminus while the zinc finger domain of subunit 3 plays a role in its binding to the 15S U2 snRNP.This This gene encodes subunit 3 of the splicing factor 3a protein complex. The splicing factor 3a heterotrimer includes subunits 1, 2 and 3 and is necessary for the in vitro conversion of 15SMore Product Information