Tisk…
Anti-XPO1 Rabbit Polyclonal Antibody
Dodavatel: ProSci Inc.
29-311
PRSI29-311EA 18800 CZK
PRSI29-311
Anti-XPO1 Rabbit Polyclonal Antibody
Protilátky
XPO1 mediates leucine-rich nuclear export signal (NES)-dependent protein transport. Exportin 1 specifically inhibits the nuclear export of Rev and U snRNAs. It is involved in the control of several cellular processes by controlling the localization of cyclin B, MPAK, and MAPKAP kinase 2. XPO1 also regulates NFAT and AP-1.The protein encoded by this gene mediates leucine-rich nuclear export signal (NES)-dependent protein transport. Exportin 1 specifically inhibits the nuclear export of Rev and U snRNAs. It is involved in the control of several cellular processes by controlling theMore Product Information
Objednat
Produktová specifikace
Symbol antigenu XPO1
Název antigenu Exportin 1
Environmentally Preferable
Synonyma antigenu Exp1|Exportin-1|Chromosome region maintenance 1 protein homolog|CRM1|XPO1
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Izotyp
Western blot Yes
ELISA Yes
Imunogen A synthetic peptide corresponding a region of human XPO1.
Formulář Lyophilized