Tisk…

Anti-POU2F3 Rabbit Polyclonal Antibody

Dodavatel: ProSci Inc.
27-625
PRSI27-625EA 18800 CZK
PRSI27-625
Anti-POU2F3 Rabbit Polyclonal Antibody
Protilátky
POU domain genes encode a family of highly conserved transacting factors that influence the transcriptional activity of several cell type-specific and ubiquitous genes.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: POU2F3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR