Tisk…
Anti-FAM219A Rabbit Polyclonal Antibody
Dodavatel: ProSci Inc.
27-034
PRSI27-034EA 18800 CZK
PRSI27-034
Anti-FAM219A Rabbit Polyclonal Antibody
Protilátky
The function of this protein has not been determined.The protein encoded by this gene has homologs that have been identified in mouse and macaque. The mouse and human proteins have a putative prenyl group binding site (CAAX box) at their C-terminus. A diverse list of proteins are known or strongly presumed to be the target of post-translational modification by the attachment of either a farnesyl or a geranyl-geranyl group to a cysteine residue at the C-terminus. The function of this protein has not been determined.
Antibody is purified by peptide affinity chromatography method.