Tisk…

Anti-GCA Rabbit Polyclonal Antibody

Dodavatel: ProSci Inc.
22-686
PRSI22-686EA 11930 CZK
PRSI22-686
Anti-GCA Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GCA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR