Tisk…

Anti-RHOD Rabbit Polyclonal Antibody

Dodavatel: ProSci Inc.
22-243
PRSI22-243EA 11930 CZK
PRSI22-243
Anti-RHOD Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: RHOD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR