Tisk…

Anti-LSM10 Rabbit Polyclonal Antibody

Dodavatel: ProSci Inc.
16-667
PRSI16-667EA 11930 CZK
PRSI16-667
Anti-LSM10 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: LSM10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
New Product
Objednat

Produktová specifikace

Symbol antigenu LSM10
Název antigenu LSM10, U7 small nuclear RNA associated
Environmentally Preferable
Synonyma antigenu U7 snRNA-associated Sm-like protein LSm1U7 snRNP-specific Sm-like protein LSM10|MST074|MSTP074
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Rat
Hostitel Rabbit
Identifikační číslo genu 84967
Izotyp IgG
Western blot Yes
Modifikace None
Imunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-123 of human LSM10 (NP_116270,1)
Formulář Liquid
Čištění Affinity purification
Koncentrace Batch dependent
Skladovací pufr PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
Skladovací teplota Store at −20 °C. Avoid freeze/thaw cycles.
Transportní teplota 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR