Tisk…

Anti-ASB13 Rabbit Polyclonal Antibody

Dodavatel: ProSci Inc.
16-309
PRSI16-309EA 11930 CZK
PRSI16-309
Anti-ASB13 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ASB13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR