Tisk…

Anti-CYP3A4 Rabbit Polyclonal Antibody

Dodavatel: ProSci Inc.
15-013
PRSI15-013EA 11930 CZK
PRSI15-013
Anti-CYP3A4 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CYP3A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR