Tisk…
Liver Lysate (0 Days Old)
Dodavatel: ProSci Inc.
1404-0
PRSI1404-0EA 4360 CZK
PRSI1404-0
Liver Lysate (0 Days Old)
Buňky
Liver tissue lysate (0 Days Old) was prepared by homogenisation in modified RIPA buffer (150 mM sodium chloride, 50 mM Tris-HCl, pH 7,4, 1 mM ethylenediaminetetraacetic acid, 1 mM phenylmethylsulfonyl fluoride, 1% Triton X-100, 1% sodium deoxycholic acid, 0,1% sodium dodecylsulfate, 5 μg/ml of aprotinin, 5 μg/ml of leupeptin.
Tissue and cell debris was removed by centrifugation. Protein concentration was determined with Bio-Rad protein assay. The product was boiled for 5 min in 1 x SDS sample buffer (50 mM Tris-HCl pH 6,8, 12,5% glycerol, 1% sodium dodecylsulfate, 0,01% bromophenol blue) containing 5% β-mercaptoethanol.
New Product
Objednat
Produktová specifikace
Environmentally Preferable
Koncentrace 2 mg/ml
Testované aplikace WB
Skladovací pufr Liver lysate (0 Days Old) is supplied in SDS sample buffer containing 5% β-mercaptoethanol
Podmínky uskladnění Store at 2...8 °C for continuous use. For extended storage, freeze working aliquots at −70 °C.