Tisk…
Terbium standard solution, for ICP, in HNO₃, CRM, VHG
Dodavatel: LGC Standards PROMOCHEM
VHG-PTBN-100 VHG-LTBN-100 VHG-PTBN-500 VHG-TTBN-100 VHG-TTBN-500
PROOVHG-PTBN-100EA 1940 CZK
PROOVHG-PTBN-100 PROOVHG-LTBN-100 PROOVHG-PTBN-500 PROOVHG-TTBN-100 PROOVHG-TTBN-500
Terbium standard solution, for ICP, in HNO₃, CRM, VHG
Standardy Standardy kationické Standardy - terbium
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifikace: Traceable to NIST, SRM.