Tisk…
Nickel standard solution, for ICP, in 5% HNO₃, VHG
Dodavatel: LGC Standards PROMOCHEM
VHG-PNIN-100 VHG-PNIN-500 VHG-PNIN-50 VHG-TNIN-100 VHG-TNIN-500
PROOVHG-PNIN-100EA 1640 CZK
PROOVHG-PNIN-100 PROOVHG-PNIN-500 PROOVHG-PNIN-50 PROOVHG-TNIN-100 PROOVHG-TNIN-500
Nickel standard solution, for ICP, in 5% HNO₃, VHG
Standardy Standardy kationické Standardy - nikl
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifikace: Traceable to NIST, SRM.