Tisk…
Acetate standard, ion chromatography CRM, VHG
Dodavatel: LGC Standards PROMOCHEM
VHG-IACET-100 VHG-IACET-500
PROOVHG-IACET-100EA 1910 CZK
PROOVHG-IACET-100 PROOVHG-IACET-500
Acetate standard, ion chromatography CRM, VHG
Standardy Standardy anionické Standardy - acetátát
This solution is intended for use as a certified reference material (CRM) and calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.
Certifikace: Traceable to NIST and CRM.