Tisk…

Florisil® for chromatography

Dodavatel: LGC Standards PROMOCHEM

SC-4181-B005
PROOSC-4181-B005EA 5250 CZK
PROOSC-4181-B005
Florisil® for chromatography
Magnesium silicate
Vzorec: MgO₃Si
MDL Number: MFCD00078249
CAS číslo: 1343-88-0
EINECS: 215-681-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR