Tisk…

(1α,2α,3β,4α,5α,6β)-1,2,3,4,5,6-Hexachlorcyklohexan

Dodavatel: LGC Standards PROMOCHEM
Nebezpečí

NMIAP1332
PROONMIAP1332EA 8170 CZK
PROONMIAP1332
(1α,2α,3β,4α,5α,6β)-1,2,3,4,5,6-Hexachlorcyklohexan
(1α,2α,3β,4α,5α,6β)-1,2,3,4,5,6-Hexachlorcyklohexan
Vzorec: C₆H₆Cl₆
Storage Temperature: Ambient
MDL Number: MFCD00135947
CAS číslo: 58-89-9
EINECS: 200-401-2
UN: 2761
ADR: 6.1,I

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR