Tisk…
Ferromanganese, high carbon, alloy, NIST-68C, reference standard
Dodavatel: LGC Standards PROMOCHEM
NIST-68C
PROONIST-68CEA 26650 CZK
PROONIST-68C
Ferromanganese, high carbon, alloy, NIST-68C, reference standard
Standardy Slitiny a kovové standardy
This standard reference material is in the powder form intended for use in checking chemical methods of analysis and in calibration with instrumental methods of analysis.
  • 8 analytes/elements measured
Certifikace: Traceable to NIST and SRM.