Tisk…
Tomato leaves - trace elements, NIST-1573A
Dodavatel: LGC Standards PROMOCHEM
NIST-1573A
PROONIST-1573AEA 52440 CZK
PROONIST-1573A
Tomato leaves - trace elements, NIST-1573A
Standardy Standardy potravinářské
This standard material is intended for use in evaluating the reliability of analytical methods for the determination of major, minor, and trace elements in botanical materials, agricultural food products, and materials of similar matrix.
A unit consists of 50 g of dried tomato leaves.
Certifikace: Traceable to NIST and SRM numbers.