Tisk…

Benzilic acid

Dodavatel: LGC Standards PROMOCHEM

MM0783.09
PROOMM0783.09EA 26060 CZK
PROOMM0783.09
Benzilic acid
Benzilic acid
Vzorec: C₁₄H₁₂O₃
Bod varu: 409 °C (1013 hPa)
Bod tání: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00004447
CAS číslo: 76-93-7
EINECS: 200-993-2
Merck Index: 13,01080

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR