Tisk…

L(+)-Arginin

Dodavatel: LGC Standards PROMOCHEM
Varování

Synonyms: H-L-Arg-OH, (S)-2-Amino-5-guanidinopentanoic acid, Arginine, (S)-(+)-Arginine, L-Arginine, H-Arg-OH

MM0568.00
PROOMM0568.00EA 39330 CZK
PROOMM0568.00
L(+)-Arginin
L(+)-Arginin
Vzorec: C₆H₁₄N₄O₂
MW: 174.2
Bod varu: 368
Bod tání: 238
Hustota: 1.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002635
CAS číslo: 74-79-3
EINECS: 200-811-1
Merck Index: 13,00785

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR