Tisk…

4'-Ethoxyacetanilide

Dodavatel: LGC Standards PROMOCHEM

ICRS0380
PROOICRS0380EA 7380 CZK
PROOICRS0380
4'-Ethoxyacetanilide
4'-Ethoxyacetanilide
Vzorec: C₁₀H₁₃NO₂
MDL Number: MFCD00009094
CAS číslo: 62-44-2
EINECS: 200-533-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR