Tisk…

2,2',3,4-Tetrachlorobiphenyl (PCB No. 41)

Dodavatel: LGC Standards PROMOCHEM

CIL-PCB-41-C
PROOCIL-PCB-41-CEA 22190 CZK
PROOCIL-PCB-41-C
2,2',3,4-Tetrachlorobiphenyl (PCB No. 41)
2,2',3,4-Tetrachlorobiphenyl (PCB No. 41)
Vzorec: C₁₂H₆Cl₄
MW: 291.99
CAS číslo: 52663-59-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR