Tisk…

2,2',3,4',5,6'-Hexachlorobiphenyl (PCB No. 148)

Dodavatel: LGC Standards PROMOCHEM

CIL-PCB-148-C
PROOCIL-PCB-148-CEA 23370 CZK
PROOCIL-PCB-148-C
2,2',3,4',5,6'-Hexachlorobiphenyl (PCB No. 148)
2,2',3,4',5,6'-Hexachlorobiphenyl (PCB No. 148)
Vzorec: C₁₂H₄Cl₆
CAS číslo: 74472-41-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR