Tisk…

2,3,4',5,6-Pentachlorobiphenyl (PCB No. 117)

Dodavatel: LGC Standards PROMOCHEM

CIL-PCB-117-C
PROOCIL-PCB-117-CEA 22190 CZK
PROOCIL-PCB-117-C
2,3,4',5,6-Pentachlorobiphenyl (PCB No. 117)
2,3,4',5,6-Pentachlorobiphenyl (PCB No. 117)
Vzorec: C₁₂H₅Cl₅
MW: 326.44
CAS číslo: 68194-11-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR