Tisk…

2,3,3',5',6-Pentachlorobiphenyl (PCB No. 113)

Dodavatel: LGC Standards PROMOCHEM

CIL-PCB-113-C
PROOCIL-PCB-113-CEA 25110 CZK
PROOCIL-PCB-113-C
2,3,3',5',6-Pentachlorobiphenyl (PCB No. 113)
2,3,3',5',6-Pentachlorobiphenyl (PCB No. 113)
Vzorec: C₁₂H₅Cl₅
CAS číslo: 68194-10-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR