Tisk…

Pilocarpine nitrate

Dodavatel: LGC Standards PROMOCHEM

BP503
PROOBP503EA 9740 CZK
PROOBP503
Pilocarpine nitrate
Pilocarpine nitrate
Vzorec: C₁₁H₁₇N₃O₅
MDL Number: MFCD00078497
CAS číslo: 148-72-1
EINECS: 205-723-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR