Tisk…
Autoclaves, medium capacity, Priorclave
Dodavatel: Priorclave
EH100-SMART EH150-SMART EH200-SMART EH320-SMART EV100-SMART EV150-SMART EH200-BASE EV150-BASE EH100-BASE EH150-BASE EV100-BASE EH320-BASE
PRIOEH100-SMARTEA 632710 CZK
PRIOEH100-SMART PRIOEH150-SMART PRIOEH200-SMART PRIOEH320-SMART PRIOEV100-SMART PRIOEV150-SMART 481-0915 481-0918 481-0913 481-0914 481-0917 481-0916
Autoclaves, medium capacity, Priorclave
Sterilizátory Autoklávy
These versatile autoclaves are fully equipped for use in laboratories throughout the food, drink, dairy, health and pharmaceutical industries and offer a wide range of options and accessories to further improve their performance to suit individual load types. This flexibility is enhanced by the TACTROL™ microprocessor control system, specially developed for laboratory autoclaves, with progamming ranging from simple cycles to fully featured multi-program operation with logged records. All Priorclaves come complete with Biomaster antimicrobial coating, helping improved laboratory hygiene; itMore Product Information