Tisk…
Swabs, PurSwab®
Dodavatel: PURITAN MEDICAL
870-PCDBL
PMPC870-PCDBLEA 12200 CZK
PMPC870-PCDBL
Swabs, PurSwab®
Tampóny odběrové Tampóny pro čištění
These double-ended swabs with cotton tips are ideal for cleaning sensitive components in critical environments.
  • Low in NVR
  • Highly absorbent and does not swell in solvents
  • Lint- and residue-free
  • Sharp, double pointed tip with fine paper handle
  • Anti-static packaging contains no silicone
Upozornení: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904