Tisk…
Imject™ Bovine serum albumin (BSA), maleimide activated
Dodavatel: Thermo Fisher Scientific
77115 77116
PIER77115EA 16980 CZK
PIER77115 PIER77116
Imject™ Bovine serum albumin (BSA), maleimide activated
Proteiny
Imject™ BSA maleimide-activated enables conjugation of sulfhydryl-containing peptide antigens to immunogenic bovine serum albumin carrier protein for immunisation and antibody production.
Objednat
Produktová specifikace
Název proteinu/peptidu Bovine Serum Albumin
Environmentally Preferable
Proteinová synonyma BSA|Fraction V|Cohn Fraction V
Druhy Bovine
Zdroj Serum