Tisk…
Desalting columns, Pierce™
Dodavatel: Thermo Fisher Scientific
43230 43233
PIER43230EA 4910 CZK
PIER43230 PIER43233
Desalting columns, Pierce™
Kolony chromatografické
These desalting columns are ready-to-use, disposable, gel filtration columns for separating proteins and other macromolecules from low molecular weight buffer salts and reagents.
  • Stable in water, salt solutions, organic solvents and alkaline or acidic conditions
  • Excellent flow properties
  • Heat stable
  • Gravity flow column format
  • Purification targeting buffer exchange, protein
  • Disposable
They have wide range of applications that includes removing salts from protein solutions, removing phenol from nucleic acid preparations, separating excess crosslinkerMore Product Information