Tisk…
Pierce™ Agarose ChIP Kit
Dodavatel: Thermo Fisher Scientific
26156
PIER26156EA 23720 CZK
PIER26156
Pierce™ Agarose ChIP Kit
Systémy pro purifikaci proteinů Protein Purification Kits
Provides a complete set of reagents and a simple, fast and reproducible protocol to perform chromatin immunoprecipitation (ChIP) assays.
  • High-recovery DNA purification
  • Highly efficient isolation and lysis of nuclei
  • Easy and reproducible enzymatic digestion
  • Low-background and high-binding capacity Protein A/G agarose resin
  • Highly specific positive controls included: RNA polymerase II antibody and GAPDH PCR primers
ChIP assay has the ability to capture a snapshot of specific protein-DNA interactions as they occur in living cells, and then quantitate the interactions using standard or quantitive PCR.