Tisk…

BMOE (bis-Maleimidoethane), Pierce™

Dodavatel: Thermo Fisher Scientific

Synonyms: Ethylene-bis-maleimide, 1,2-Bis-maleimidoethane, 1,2-Bismaleimidoethane, 1,2-Di(maleimido)ethane, N,N'-Ethylenebismaleimide, 1,2-Bis(maleimido)ethane, 1,1'-(Ethane-1,2-diyl)bis(1H-pyrrole-2,5-dione), BMOE, N,N'-Ethylenedimaleimide

22323
PIER22323EA 5210 CZK
PIER22323
BMOE (bis-Maleimidoethane), Pierce™
BMOE (bis-Maleimidoethane)

Thermo Scientific Pierce BMOE is a short-arm, maleimide crosslinker for covalent, irreversible conjugation between sulfhydryl groups (e.g., protein or peptide cysteines).


  • Reactive groups: maleimide (both ends)
  • Reactive towards: sulfhydryl groups
  • Short (8.0 Å), sulfhydryl-to-sulfhydryl crosslinker, composed of maleimide groups and 7-atom spacer arm
  • Shortest bismaleimide crosslinker available for close proximity crosslinking
  • Water-insoluble—dissolve first in DMF or DMSO, then add to aqueous reaction buffers
  • Noncleavable


Upozornení: For Research Use Only. Not for use in diagnostic procedures.

Vzorec: C₁₀H₈N₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD00458699
CAS číslo: 5132-30-9

Objednat

Výsledky specifikačního testu

Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety Maleimide
Spacer Short
Form Solid
Quantity 50 mg
Type Crosslinker
Solubility DMF, DMSO
Spacer Arm 8.0 Å


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR