Tisk…

D(+)-Glukóza, Supelco®

Dodavatel: Merck

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

PHR1000-1G
PHR1000-1GEA 3070 CZK
PHR1000-1G
D(+)-Glukóza, Supelco®
D(+)-Glukóza
Vzorec: C₆H₁₂O₆
Bod tání: 146 °C
Hustota: 1,54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00148912
CAS číslo: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f49f5d1cb18b453e46cea2b1746a654914e917e&vid=2f49f5d1cb18b453e46cea2b1746a654914e917e&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR