Tisk…

Foaming Solutions for Hygienic Hand Washing

Dodavatel: PHAGOGENE
65123 SA0141050
PHAG65123EA 1970 CZK
PHAG65123 PHAGSA0141050
Foaming Solutions for Hygienic Hand Washing
Péče o pokožku Mýdla a čističe pokožky
Liquid washing and disinfecting soap specially formulated for the hygienic hands washing.

  • Skin-friendly formula even in case of frequent washing
  • Leave your hands soft
  • Enriched with glycerine
  • Without perfume

Used before any aseptic gesture (catheters and more) and systematically before and after any contact with a patient or contaminating material. Specially formulated for the medical, food and institutional sectors.

Appearance: Gelled liquid, opalescent
Colour: Colourless
Odor: Alcohol
pH at 20 °C (pure product): 7,0 ±0,5
Density at 20 °C (g/ml): 0,905More Product Information
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR